5-Methyl-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate 98+%

Supplier: Ambeed
Danger

A1462563-1G A1462563-250MG A1462563-5G
76662-094EA 68 USD
76662-094 77597-316 77874-983
5-Methyl-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate 98+%
5-Methyl-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O₃S₂
CAS Number: 112359-25-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR