Methyl 5-(bis((1,1-dimethylethoxy)carbonyl)amino)-2-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

A1462507-100MG A1462507-1G A1462507-250MG A1462507-50MG
76662-086EA 128.37 USD
76662-086 76662-088 76662-090 76662-092
Methyl 5-(bis((1,1-dimethylethoxy)carbonyl)amino)-2-pyridinecarboxylate 97%
Methyl 5-(bis((1,1-dimethylethoxy)carbonyl)amino)-2-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₆
MW: 352.3823 g/mol
CAS Number: 2243961-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR