Guanosine 5'-triphosphate disodium salt ≥93% (contains < 7% H₂O)

Supplier: Ambeed
Warning

Synonyms: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt

A1462463-250MG A1462463-25G A1462463-1G A1462463-5G
76662-080EA 18.04 USD
76662-080 77600-092 77597-312 77597-314
Guanosine 5'-triphosphate disodium salt ≥93% (contains < 7% H₂O)
Guanosine 5'-triphosphate disodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₁₄P₃
MW: 567.147 g/mol
MDL Number: MFCD03410297
CAS Number: 56001-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR