6,6'-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥97%, ee 99%

Supplier: Ambeed
Warning

A1462424-100MG A1462424-250MG
76662-076EA 155 USD
76662-076 76662-078
6,6'-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine ≥97%, ee 99%
6,6'-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₂₈H₂₂N₄O₂
MW: 446.4999 g/mol
CAS Number: 2757082-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR