6-(2-(Methylsulfonyl)pyrimidin-5-yl)hex-5-ynoic acid 97%

Supplier: Ambeed
Warning

A1462228-100MG A1462228-250MG A1462228-5G
76662-056EA 63.66 USD
76662-056 76662-058 76662-060
6-(2-(Methylsulfonyl)pyrimidin-5-yl)hex-5-ynoic acid 97%
6-(2-(Methylsulfonyl)pyrimidin-5-yl)hex-5-ynoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₄S
MW: 268.2890 g/mol
MDL Number: MFCD34186943
CAS Number: 2356229-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR