4-Bromo-3-(trifluoromethoxy)pyridine hydrobromide 97%

Supplier: Ambeed
Warning

A1462193-1G A1462193-250MG
76662-050EA 622.75 USD
76662-050 76662-052
4-Bromo-3-(trifluoromethoxy)pyridine hydrobromide 97%
4-Bromo-3-(trifluoromethoxy)pyridine hydrobromide

Precaución: for Research Use Only

Formula: C₆H₄Br₂F₃NO
MW: 322.9053 g/mol
MDL Number: MFCD34178761
CAS Number: 2665660-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR