Dibromobis(t-Bu pyridine)nickel 98%

Supplier: Ambeed
Danger

A1461982-1G A1461982-25G A1461982-5G
76662-042EA 91 USD
76662-042 77874-975 77874-976
Dibromobis(t-Bu pyridine)nickel 98%
Dibromobis(t-Bu pyridine)nickel

Precaución: for Research Use Only

Formula: C₁₈H₂₆Br₂N₂Ni
CAS Number: 2640505-89-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR