6-Amino-4-fluoropyridin-3-ol hydrochloride 97%

Supplier: Ambeed
Warning

A1461956-100MG A1461956-250MG A1461956-1G
76662-036EA 178 USD
76662-036 76662-038 77624-222
6-Amino-4-fluoropyridin-3-ol hydrochloride 97%
6-Amino-4-fluoropyridin-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₆ClFN₂O
MDL Number: MFCD34178760
CAS Number: 2703752-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR