(4R,4'R)-2,2'-(1,3-Di(quinolin-2-yl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1461807-100MG A1461807-1G A1461807-250MG
76662-022EA 128.37 USD
76662-022 76662-024 76662-026
(4R,4'R)-2,2'-(1,3-Di(quinolin-2-yl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) ≥97%, ee 99%
(4R,4'R)-2,2'-(1,3-Di(quinolin-2-yl)propane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₄₁H₃₆N₄O₂
MW: 616.7500 g/mol
CAS Number: 2828433-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR