(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A1461789-100MG A1461789-1G A1461789-250MG A1461789-5G
76662-014EA 79.57 USD
76662-014 76662-016 76662-018 76662-020
(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%
(1R,2R)-2-Aminocyclopentanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.6180 g/mol
MDL Number: MFCD09749901
CAS Number: 158414-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR