2-(4-Phenoxyphenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1461194-1G A1461194-250MG A1461194-5G
76661-990EA 26.53 USD
76661-990 76661-992 76661-994
2-(4-Phenoxyphenyl)-1,3,2-dioxaborinane 98%
2-(4-Phenoxyphenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₅H₁₅BO₃
MW: 254.0888 g/mol
CAS Number: 2222401-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR