2-(3-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%
Supplier: Ambeed
|
Warning
|
A1461191-1G
A1461191-5G
76661-976EA
26.53
USD
76661-976
76661-978
2-(3-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%
2-(3-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane
Precaución: for Research Use Only
Formula:
C₁₀H₁₀BF₃O₂ MW: 229.9914 g/mol |
CAS Number:
2070921-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...