2-(2-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1461190-250MG A1461190-5G
76661-972EA 10.61 USD
76661-972 76661-974
2-(2-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane 98%
2-(2-(Trifluoromethyl)phenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₀H₁₀BF₃O₂
MW: 229.9914 g/mol
CAS Number: 1400664-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR