2-(4-(tert-Butyl)phenyl)-1,3,2-dioxaborinane 98%

Supplier: Ambeed
Warning

A1461188-1G A1461188-250MG A1461188-5G
76661-966EA 25 USD
76661-966 76661-968 76661-970
2-(4-(tert-Butyl)phenyl)-1,3,2-dioxaborinane 98%
2-(4-(tert-Butyl)phenyl)-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₂
CAS Number: 1370412-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR