4-(4-(Trifluoromethyl)phenyl)pyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A1461077-100MG A1461077-1G A1461077-250MG
76661-946EA 128.37 USD
76661-946 76661-948 76661-950
4-(4-(Trifluoromethyl)phenyl)pyrimidin-2-amine 97%
4-(4-(Trifluoromethyl)phenyl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₈F₃N₃
MW: 239.2000 g/mol
MDL Number: MFCD07381878
CAS Number: 916792-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR