6-(4-(Trifluoromethyl)phenyl)pyrazin-2-amine 97%

Supplier: Ambeed
Warning

A1461075-100MG A1461075-1G A1461075-250MG
76661-940EA 98.67 USD
76661-940 76661-942 76661-944
6-(4-(Trifluoromethyl)phenyl)pyrazin-2-amine 97%
6-(4-(Trifluoromethyl)phenyl)pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₈F₃N₃
MW: 239.2000 g/mol
MDL Number: MFCD12033131
CAS Number: 1159819-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR