6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A1461073-1G A1461073-250MG
76661-936EA 583.5 USD
76661-936 76661-938
6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine 97%
6-(4-(Trifluoromethyl)phenyl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₁H₈F₃N₃
MW: 239.2000 g/mol
MDL Number: MFCD20838702
CAS Number: 1491938-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR