Acid red 27 85%

Supplier: Ambeed
Warning

A1460430-1000G A1460430-100G A1460430-25G A1460430-500G
76661-916EA 87 USD
76661-916 76661-918 76661-920 76661-922
Acid red 27 85%
Acid red 27

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O₁₀S₃
MDL Number: MFCD00004076
CAS Number: 915-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR