Pyridine-2,3-dicarboxylic acid 99%

Supplier: Ambeed
Warning

A1460048-100G A1460048-25G A1460048-500G
76661-910EA 13 USD
76661-910 76661-912 76661-914
Pyridine-2,3-dicarboxylic acid 99%
Pyridine-2,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₄
MDL Number: MFCD00006295
CAS Number: 89-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR