Aluminum(III) ammonium sulfate dodecahydrate GR

Supplier: Ambeed
Warning

A1459586-1000G A1459586-500G
76661-896EA 15 USD
76661-896 76661-898
Aluminum(III) ammonium sulfate dodecahydrate GR
Aluminum(III) ammonium sulfate dodecahydrate

Precaución: for Research Use Only

Formula: AlH₂₈NO₂₀S₂
MDL Number: MFCD00149958
CAS Number: 7784-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR