Ammonium iron (II) sulfate hexahydrate 99.8%, Analytical reagent (AR)

Supplier: Ambeed
Warning

Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate

A1459579-1000G A1459579-25G A1459579-500G A1459579-100G
76661-892EA 22.28 USD
76661-892 76661-894 77597-284 77624-216
Ammonium iron (II) sulfate hexahydrate 99.8%, Analytical reagent (AR)
Ammonium iron(II) sulphate hexahydrate

Precaución: for Research Use Only

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
MDL Number: MFCD00150530
CAS Number: 7783-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR