Sulfamethoxazole 99+%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide

A1458841-1000G A1458841-100G A1458841-25G A1458841-500G
76661-878EA 137.92 USD
76661-878 76661-880 76661-882 76661-884
Sulfamethoxazole 99+%
Sulfamethoxazole

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃O₃S
MW: 253.28 g/mol
MDL Number: MFCD00010546
CAS Number: 723-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR