Manganese(II) acetate ≥98% (Loss on drying ≤30%)
Supplier: Ambeed
|
Warning
|
A1458432-100G
A1458432-5G
76661-866EA
84
USD
76661-866
76661-868
Manganese(II) acetate ≥98% (Loss on drying ≤30%)
Manganese(II) acetate
Precaución: for Research Use Only
Formula:
(H₃CCOO)₂Mn |
MDL Number:
MFCD00013039 CAS Number: 638-38-0 |
Specification Test Results
Loss on drying | ≤ 30% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...