Bromocresol green sodium salt indicator

Supplier: Ambeed
Warning

A1458250-100G A1458250-25G A1458250-5G
76661-852EA 226 USD
76661-852 76661-854 76661-856
Bromocresol green sodium salt indicator
Bromocresol green sodium salt

Precaución: for Research Use Only

Formula: C₂₁H₁₃Br₄NaO₅S
MDL Number: MFCD00148898
CAS Number: 62625-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR