Leuco crystal violet 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)

A1458078-1000G A1458078-100G A1458078-25G A1458078-500G A1458078-5G
76661-834EA 121 USD
76661-834 76661-836 76661-838 76661-840 76661-842
Leuco crystal violet 98% (by HPLC)
Leuco crystal violet

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
MDL Number: MFCD00008314
CAS Number: 603-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR