Pararosaniline hydrochloride > 85 % (dye content)

Supplier: Ambeed
Danger

A1457769-25G A1457769-5G
76661-828EA 24 USD
76661-828 76661-830
Pararosaniline hydrochloride > 85 % (dye content)
Pararosaniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₁₈ClN₃
MDL Number: MFCD00001657
CAS Number: 569-61-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR