Sodium-2-naphthalenesulfonate ≥99.0% for ion pair chromatography
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Naphthalenesulphonic acid Sodium Salt, Sodium 2-naphthalenesulphonate
A1457444-1000G
A1457444-100G
A1457444-25G
A1457444-500G
76661-816EA
138.98
USD
76661-816
76661-818
76661-820
76661-822
Sodium-2-naphthalenesulfonate ≥99.0% for ion pair chromatography
Sodium-2-naphthalenesulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₈O₃S MW: 230.21 g/mol |
MDL Number:
MFCD00064186 CAS Number: 532-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...