Rhodizonic acid disodium salt, Analytical reagent (AR)

Supplier: Ambeed
Warning

Synonyms: Sodium rhodizonate

A1457365-100G A1457365-25G A1457365-5G
76661-804EA 88.06 USD
76661-804 76661-806 76661-808
Rhodizonic acid disodium salt, Analytical reagent (AR)
Rhodizonic acid disodium salt

Precaución: for Research Use Only

Formula: C₆Na₂O₆
MW: 214.04 g/mol
MDL Number: MFCD00001650
CAS Number: 523-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR