3,5-Dichlorobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A1457284-1000G
A1457284-100G
A1457284-25G
A1457284-500G
76661-796EA
99.73
USD
76661-796
76661-798
76661-800
76661-802
3,5-Dichlorobenzoic acid 95%
3,5-Dichlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol |
MDL Number:
MFCD00002494 CAS Number: 51-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...