Methyl red acid-base indicator 4.5 (red) - 6.2 (yellow)
Supplier: Ambeed
|
Warning
|
A1457118-25G
A1457118-1000G
A1457118-100G
76661-794EA
20
USD
76661-794
77874-948
77874-949
Methyl red acid-base indicator 4.5 (red) - 6.2 (yellow)
Methyl red
Precaución: for Research Use Only
Formula:
C₁₅H₁₅N₃O₂ |
MDL Number:
MFCD00002425 CAS Number: 493-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...