DL-Sulforaphane 99%

Supplier: Ambeed
Danger

A1456978-100MG A1456978-1G A1456978-1MG A1456978-250MG A1456978-5MG
76661-790EA 235 USD
76661-790 76661-792 77622-182 77622-184 77874-947
DL-Sulforaphane 99%
DL-Sulforaphane

Precaución: for Research Use Only

Formula: C₆H₁₁NOS₂
MDL Number: MFCD00198068
CAS Number: 4478-93-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR