2-Oxoglutaric acid 98% (HPLC-ELSD)

Supplier: Ambeed
Danger

Synonyms: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid

A1456545-1000G A1456545-100G A1456545-25G A1456545-500G
76661-770EA 103.97 USD
76661-770 76661-772 76661-774 76661-776
2-Oxoglutaric acid 98% (HPLC-ELSD)
2-Oxoglutaric acid

Precaución: for Research Use Only

Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
MDL Number: MFCD00004165
CAS Number: 328-50-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR