Polyethylene glycol 3,350 ∼ Mn 3350

Supplier: Ambeed
Warning

Synonyms: PEG 3,350, PEG 3350

A1456080-500G A1456080-100G
76661-764EA 11 USD
76661-764 77597-266
Polyethylene glycol 3,350 ∼ Mn 3350
Polyethylene glycol 3,350

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR