Tris(8-quinolinolato)aluminum 99.9%
Supplier: Ambeed
|
Warning
|
Synonyms:
8-Quinolinol Aluminum Complex, Alq3, Aluminum Tris(8-hydroxyquinolinate), Aluminum triquinolin-8-olate
A1455784-100G
A1455784-1G
A1455784-25G
76661-746EA
262
USD
76661-746
76661-748
76661-750
Tris(8-quinolinolato)aluminum 99.9%
Tris(8-quinolinolato)aluminum
Precaución: for Research Use Only
Formula:
C₂₇H₁₈AlN₃O₃ MW: 459.44 g/mol |
MDL Number:
MFCD00191693 CAS Number: 2085-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...