2-tert-Butylhydroquinone ≥98%

Supplier: Ambeed
Warning

Synonyms: t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ

A1455700-100G A1455700-25G A1455700-500G
76661-740EA 9.55 USD
76661-740 76661-742 76661-744
2-tert-Butylhydroquinone ≥98%
2-tert-Butylhydroquinone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00002344
CAS Number: 1948-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR