Phloxin B ≥90% (dye content)
Supplier: Ambeed
|
Danger
|
A1455591-25G
A1455591-5G
A1455591-100G
76661-730EA
21.22
USD
76661-730
76661-732
77622-176
Phloxin B ≥90% (dye content)
Phloxin B
Precaución: for Research Use Only
Formula:
C₂₀H₂Br₄Cl₄Na₂O₅ |
MDL Number:
MFCD00070627 CAS Number: 18472-87-2 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...