Erythrosin B disodium salt 85%

Supplier: Ambeed
Warning

Synonyms: C.I. 45430

A1455392-100G A1455392-25G A1455392-5G
76661-716EA 58.35 USD
76661-716 76661-718 76661-720
Erythrosin B disodium salt 85%
Erythrosin B disodium salt

Precaución: for Research Use Only

Formula: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
MDL Number: MFCD00144257
CAS Number: 16423-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR