Xylenol orange triturate indicator

Supplier: Ambeed
Warning

Synonyms: 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid

A1455374-100G A1455374-25G
76661-714EA 77 USD
76661-714 77622-174
Xylenol orange triturate indicator
Xylenol orange triturate

Precaución: for Research Use Only

Formula: C₃₁H₃₂N₂O₁₃S
MDL Number: MFCD00014580
CAS Number: 1611-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR