Pefloxacine methanesulfonate dihydrate 99.5%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid mesylate dihydrate, Pefloxacin mesylate dihydrate
A1455244-100G
A1455244-25G
76661-710EA
169.75
USD
76661-710
76661-712
Pefloxacine methanesulfonate dihydrate 99.5%
Pefloxacine methanesulfonate dihydrate
Precaución: for Research Use Only
Formula:
C₁₇H₂₀FN₃O₃·CH₄O₃S·2H₂O MW: 333.4 · 96.1 · 36.0 g/mol |
MDL Number:
MFCD01685696 CAS Number: 149676-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...