Nickel diiodide 99%

Supplier: Ambeed
Danger

Synonyms: Nickel (II) iodide, Nickel iodide (NiI2)

A1454837-25G A1454837-5G A1454837-1G A1454837-100G A1454837-500G
76661-686EA 27 USD
76661-686 76661-688 77597-256 77622-170 77867-816
Nickel diiodide 99%
Nickel diiodide

Precaución: for Research Use Only

Formula: I₂Ni
MDL Number: MFCD00016264
CAS Number: 13462-90-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR