(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate 96%

Supplier: Ambeed
Warning

A1454705-1G A1454705-250MG A1454705-5G
76661-678EA 57 USD
76661-678 76661-680 76661-682
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate 96%
(S)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-(tritylamino)butanoate

Precaución: for Research Use Only

Formula: C₄₄H₃₁F₅N₂O₅
MDL Number: MFCD00153373
CAS Number: 132388-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR