Zinc sulphide 99.99% (metals basis), powder 3.3-5 μm

Supplier: Ambeed
Warning

Synonyms: Zinc (II) sulphide

A1454626-25G A1454626-500G A1454626-100G
76661-672EA 123.07 USD
76661-672 76661-674 76661-676 77597-254
Zinc sulphide 99.99% (metals basis), powder 3.3-5 μm
Zinc sulphide

Precaución: for Research Use Only

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
MDL Number: MFCD00011301
CAS Number: 1314-98-3
Merck Index: 12,10294

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR