Ammonium sodium hydrogen phosphate 99%, Analytical reagent (AR)

Supplier: Ambeed
Warning

A1454416-1000G A1454416-100G A1454416-25G A1454416-500G
76661-664EA 89 USD
76661-664 76661-666 76661-668 76661-670
Ammonium sodium hydrogen phosphate 99%, Analytical reagent (AR)
Ammonium sodium hydrogen phosphate

Precaución: for Research Use Only

Formula: H₅NNaO₄P
MDL Number: MFCD00066293
CAS Number: 13011-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR