Benzethonium chloride 99+%

Supplier: Ambeed
Warning

Synonyms: Hyamine

A1454142-1000G A1454142-100G A1454142-25G A1454142-500G
76661-650EA 120.95 USD
76661-650 76661-652 76661-654 76661-656
Benzethonium chloride 99+%
Benzethonium chloride

Precaución: for Research Use Only

Formula: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
MDL Number: MFCD00011742
CAS Number: 121-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR