2-Imidazolidinone 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Imidazolidone
A1454114-500G
A1454114-25G
A1454114-100G
76661-648EA
33.95
USD
76661-648
77566-942
77597-250
2-Imidazolidinone 99%
2-Imidazolidinone
Precaución: For research use only.
Formula:
C₃H₆N₂O MW: 86.0925 g/mol |
MDL Number:
MFCD00005257 CAS Number: 120-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...