4-Amino-3-hydroxy-1-naphthalenesulfonic acid ≥90%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
A1453894-100G
A1453894-25G
76661-642EA
55.17
USD
76661-642
76661-644
4-Amino-3-hydroxy-1-naphthalenesulfonic acid ≥90%
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₄S MW: 239.25 g/mol |
MDL Number:
MFCD00004019 CAS Number: 116-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...