Orbifloxacin 99%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-7-(cis-3,5-dimethylpiperazin-1-yl)-5,6,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

A1453840-100MG A1453840-1G A1453840-250MG A1453840-5G
76661-634EA 26 USD
76661-634 76661-636 76661-638 76661-640
Orbifloxacin 99%
Orbifloxacin

Precaución: for Research Use Only

Formula: C₁₉H₂₀F₃N₃O₃
MDL Number: MFCD00864858
CAS Number: 113617-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR