Methyl palmitate 99% (by GC)
Supplier: Ambeed
|
Warning
|
A1453733-100G
A1453733-25G
A1453733-500G
A1453733-1000G
76661-614EA
21
USD
76661-614
76661-616
76661-618
77874-927
Methyl palmitate 99% (by GC)
Methyl palmitate
Precaución: for Research Use Only
Formula:
C₁₇H₃₄O₂ |
MDL Number:
MFCD00008994 CAS Number: 112-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...