Triethylene glycol ≥99%, Analytical reagent (AR)

Supplier: Ambeed
Warning

A1453718-1000G A1453718-500G A1453718-100G
76661-608EA 15.99 USD
76661-608 76661-610 77566-940
Triethylene glycol ≥99%, Analytical reagent (AR)
Triethylene glycol

Precaución: For research use only.

Formula: C₆H₁₄O₄
MDL Number: MFCD00002880
CAS Number: 112-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR