Octylamine ≥99%

Supplier: Ambeed
Danger

Synonyms: 1-Aminooctane, n-Octylamine, 1-Octylamine

A1453661-25G A1453661-100G
76661-596EA 6.37 USD
76661-596 77622-166
Octylamine ≥99%
Octylamine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₆CH₂NH₂
MW: 129.25 g/mol
MDL Number: MFCD00008247
CAS Number: 111-86-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR